Keep my session open?
Ending In 
The session is expired
Vaša relácia uplynula. Pre Vašu bezpečnosť sme Vás odhlásili.
Chcete sa opäť prihlásiť?

Anti-NR3C1 Rabbit Polyclonal Antibody

Dodávateľ: OriGene
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100; Immunofluorescence: 1:100-1:500; ELISA: 1:40000

Type: Primary
Antigen: NR3C1 (nuclear receptor subfamily 3, group C, member 1 (glucocorticoid receptor))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat



 

OBJEDNAŤ

Rozšíriť všetko / Schovať všetko
Veľkosť Pk Číslo v katalógu VWR Množstvo
100 µl 100 µl ORIGTA313373
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

ŠPECIFIKÁCIE

Antigen symbol NR3C1
Antigen name nuclear receptor subfamily 3, group C, member 1 (glucocorticoid receptor)
Reactivity Human, Mouse, Rat
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
Cross adsorption none
Purification purified by affinity chromatography
Form Liquid
Storage buffer Phosphate buffered saline (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
Storage temperature Cool
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping