Tlačiť

Anti-ATP7B Rabbit Polyclonal Antibody

Dodávateľ: OriGene
ORIGTA313380EA 626.97 EUR
ORIGTA313380
Anti-ATP7B Rabbit Polyclonal Antibody
Protilátky
Recommended Dilutions: Immunochemistry: 1:50-1:100; Immunofluorescence: 1:100-1:500; ELISA: 1:1000

Type: Primary
Antigen: ATP7B (ATPase, Cu++ transporting, beta polypeptide)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR