Tlačiť

Anti-CXCL13 Mouse Polyclonal Antibody

ABNOH00010563-B01
Anti-CXCL13 Mouse Polyclonal Antibody
Type: Primary
Antigen: CXCL13 (chemokine (C-X-C motif) ligand 13)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR