Tlačiť
Anti-AHSA1 Mouse Polyclonal Antibody
Dodávateľ: Abnova
ABNOH00010598-B01EA 466.46 EUR
ABNOH00010598-B01
Anti-AHSA1 Mouse Polyclonal Antibody
Protilátky
Type: Primary
Antigen: AHSA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human