Tlačiť

Anti-FAM117B Rabbit Polyclonal Antibody

Dodávateľ: Biorbyt
BIRBORB126924-100EA 0 EUR
BIRBORB126924-100 BIRBORB126924-200
Anti-FAM117B Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: FAM117B (family with sequence similarity 117, member B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR