Tlačiť

Anti-PUF60 Rabbit Polyclonal Antibody

Dodávateľ: Biorbyt
BIRBORB126925-200EA 0 EUR
BIRBORB126925-200 BIRBORB126925-100
Anti-PUF60 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: PUF60 (poly-U binding splicing factor 60KDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR