Tlačiť

Anti-FER Rabbit Polyclonal Antibody

Dodávateľ: Biorbyt
BIRBORB126928-200EA 0 EUR
BIRBORB126928-200 BIRBORB126928-100
Anti-FER Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: FER (fer (fps/fes related) tyrosine kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR