Tlačiť

Anti-FOXB1/2 Rabbit Polyclonal Antibody

Dodávateľ: Biorbyt
BIRBORB126934-100EA 0 EUR
BIRBORB126934-100 BIRBORB126934-200
Anti-FOXB1/2 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: FOXB1/2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR