Tlačiť

Anti-FOXD3 Rabbit Polyclonal Antibody

Dodávateľ: Biorbyt
BIRBORB126936-100EA 0 EUR
BIRBORB126936-100 BIRBORB126936-200
Anti-FOXD3 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: FOXD3 (forkhead box D3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR