Tlačiť

Anti-FOXH1 Rabbit Polyclonal Antibody

Dodávateľ: Biorbyt
BIRBORB126938-200EA 0 EUR
BIRBORB126938-200 BIRBORB126938-100
Anti-FOXH1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: FOXH1 (forkhead box H1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR