Tlačiť

Anti-GNAT2 Rabbit Polyclonal Antibody

Dodávateľ: Biorbyt
BIRBORB126952-200EA 0 EUR
BIRBORB126952-200 BIRBORB126952-100
Anti-GNAT2 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: GNAT2 (guanine nucleotide binding protein (G protein), alpha transducing activity polypeptide 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR