Tlačiť

Anti-GTPBP2 Rabbit Polyclonal Antibody

Dodávateľ: Biorbyt
BIRBORB126970-100EA 0 EUR
BIRBORB126970-100 BIRBORB126970-200
Anti-GTPBP2 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: GTPBP2 (GTP binding protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR