Tlačiť

Anti-Cytochrome c-type Heme Lyase Rabbit Polyclonal Antibody

Dodávateľ: Biorbyt
BIRBORB126973-200EA 0 EUR
BIRBORB126973-200 BIRBORB126973-100
Anti-Cytochrome c-type Heme Lyase Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: Cytochrome c-type Heme Lyase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR