Tlačiť

Anti-HCFC1 Rabbit Polyclonal Antibody

Dodávateľ: Biorbyt
BIRBORB126974-100EA 0 EUR
BIRBORB126974-100 BIRBORB126974-200
Anti-HCFC1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: HCFC1 (host cell factor C1 (VP16-accessory protein))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR