Tlačiť

Anti-HDAC6 Rabbit Polyclonal Antibody

Dodávateľ: Biorbyt
BIRBORB126976-100EA 0 EUR
BIRBORB126976-100 BIRBORB126976-200
Anti-HDAC6 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: HDAC6 (histone deacetylase 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR