Tlačiť

Anti-HNRNPL Rabbit Polyclonal Antibody

Dodávateľ: Biorbyt
BIRBORB126981-200EA 0 EUR
BIRBORB126981-200 BIRBORB126981-100
Anti-HNRNPL Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: HNRNPL (heterogeneous nuclear ribonucleoprotein L)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR