Tlačiť
Anti-MSI2 Rabbit Polyclonal Antibody
Dodávateľ: Abcam
ABCAAB50829-100EA 875.07 EUR
ABCAAB50829-100
Anti-MSI2 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: MSI2 (musashi RNA-binding protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now