Tlačiť

Anti-SCN5A Rabbit Polyclonal Antibody

Dodávateľ: Abcam
ABCAAB53724-100EA 1013.63 EUR
ABCAAB53724-100
Anti-SCN5A Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: SCN5A (sodium channel, voltage-gated, type V, alpha subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR