Tlačiť
Anti-THRA Rabbit Polyclonal Antibody
Dodávateľ: Abcam
ABCAAB53729-100EA 1136.93 EUR
ABCAAB53729-100
Anti-THRA Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: THRA (thyroid hormone receptor, alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat