Tlačiť

Anti-TIMP2 Rabbit Polyclonal Antibody

Dodávateľ: Abcam
ABCAAB53730-100EA 1068.17 EUR
ABCAAB53730-100
Anti-TIMP2 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: TIMP2 (TIMP metallopeptidase inhibitor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR