Tlačiť

Anti-NRIP1 Mouse Monoclonal Antibody [clone: 2656C6a]

Dodávateľ: Abcam
ABCAAB53740-100EA 1068.17 EUR
ABCAAB53740-100
Anti-NRIP1 Mouse Monoclonal Antibody [clone: 2656C6a]
Protilátky
Type: Primary
Antigen: NRIP1 (nuclear receptor interacting protein 1)
Clonality: monoclonal
Clone: 2656C6a
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR