Tlačiť
Anti-DNA polymerase delta p50 Mouse Monoclonal Antibody [clone: 2762C3a]
Dodávateľ: Abcam
ABCAAB53741-100EA 1106.86 EUR
ABCAAB53741-100
Anti-DNA polymerase delta p50 Mouse Monoclonal Antibody [clone: 2762C3a]
Protilátky
Type: Primary
Antigen: DNA polymerase delta p50
Clonality: monoclonal
Clone: 2762C3a
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human