Tlačiť

Anti-MATR3 Mouse monoclonal antibody [clone: 2539C3a]

ABCAAB53748-50
Anti-MATR3 Mouse monoclonal antibody [clone: 2539C3a]
Type: Primary
Antigen: MATR3 (matrin 3)
Clonality: monoclonal
Clone: 2539C3a
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR