Tlačiť
Anti-NUP93 Mouse Monoclonal Antibody [clone: 3332C2a]
Dodávateľ: Abcam
ABCAAB53750-100EA 1011.44 EUR
ABCAAB53750-100
Anti-NUP93 Mouse Monoclonal Antibody [clone: 3332C2a]
Protilátky
Type: Primary
Antigen: NUP93 (nucleoporin 93kDa)
Clonality: monoclonal
Clone: 3332C2a
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human