Keep my session open?
Ending In 
The session is expired
Vaša relácia uplynula. Pre Vašu bezpečnosť sme Vás odhlásili.
Chcete sa opäť prihlásiť?

Anti-DUSP4 Mouse monoclonal antibody unconjugated [clone: 2295E1a]

Dodávateľ: Abcam
Type: Primary
Antigen: DUSP4 (dual specificity phosphatase 4)
Clonality: monoclonal
Clone: 2295E1a
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:



 

OBJEDNAŤ

Rozšíriť všetko / Schovať všetko
Veľkosť Pk Číslo v katalógu VWR Množstvo
100 µg 100 µG ABCAAB53753-100
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

ŠPECIFIKÁCIE

Antigen symbol DUSP4
Antigen name Dual specificity phosphatase 4
Conjugation Unconjugated
Clonality Monoclonal
Antibody type Primary
Clone 2295E1a
Host Mouse
Isotype IgG1
Western blot Yes
Storage temperature Cool
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo