Tlačiť

Anti-ZNF81 Mouse monoclonal antibody unconjugated [clone: 3426C1a]

ABCAAB53757-100
Anti-ZNF81 Mouse monoclonal antibody unconjugated [clone: 3426C1a]
Type: Primary
Antigen: ZNF81 (zinc finger protein 81)
Clonality: monoclonal
Clone: 3426C1a
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR