Tlačiť

Anti-Prostate Specific Antigen Rabbit Polyclonal Antibody

Dodávateľ: Abcam
ABCAAB53774-100EA 1098.19 EUR
ABCAAB53774-100
Anti-Prostate Specific Antigen Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: Prostate Specific Antigen
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR