Tlačiť
Anti-RAN Rabbit Polyclonal Antibody
Dodávateľ: Abcam
ABCAAB53775-100EA 1109.27 EUR
ABCAAB53775-100
Anti-RAN Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: RAN (RAN, member RAS oncogene family)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat