Tlačiť

Anti-SYK Rabbit polyclonal antibody

ABCAAB53777-100
Anti-SYK Rabbit polyclonal antibody
Type: Primary
Antigen: SYK (spleen tyrosine kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR