Tlačiť

Anti-DP1 Mouse monoclonal antibody [clone: SPM178]

ABCAAB53924-7
Anti-DP1 Mouse monoclonal antibody [clone: SPM178]
Type: Primary
Antigen: DP1
Clonality: monoclonal
Clone: SPM178
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR