Tlačiť

Anti-Cytochrome P450 2C23 Rabbit Polyclonal Antibody

Dodávateľ: Abcam
ABCAAB53944-50EA 1112.81 EUR
ABCAAB53944-50
Anti-Cytochrome P450 2C23 Rabbit Polyclonal Antibody
Type: Primary
Antigen: Cytochrome P450 2C23
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR