Tlačiť

Anti-4-Pregnen-17, 20 beta- Diol-3-One Rabbit polyclonal antibody unconjugated

ABCAAB53950-150
Anti-4-Pregnen-17, 20 beta- Diol-3-One Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: 4-Pregnen-17, 20 beta- Diol-3-One
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR