Tlačiť

Anti-IAPP Rabbit Polyclonal Antibody

Dodávateľ: Abcam
ABCAAB53952-150EA 837.81 EUR
ABCAAB53952-150
Anti-IAPP Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: IAPP (islet amyloid polypeptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR