Keep my session open?
Ending In 
The session is expired
Vaša relácia uplynula. Pre Vašu bezpečnosť sme Vás odhlásili.
Chcete sa opäť prihlásiť?

Anti-Ly6c Rat monoclonal antibody APC (Allophycocyanin) [clone: HK1.4]

Dodávateľ: Abcam
Type: Primary
Antigen: Ly6c
Clonality: monoclonal
Clone: HK1.4
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2c
Reactivity: Mouse



 

OBJEDNAŤ

Rozšíriť všetko / Schovať všetko
Veľkosť Pk Číslo v katalógu VWR Množstvo
100 µg 100 µG ABCAAB93550-100
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

ŠPECIFIKÁCIE

Antigen symbol LY6c
Reactivity Mouse
Conjugation APC (Allophycocyanin)
Clonality Monoclonal
Antibody type Primary
Clone HK1.4
Host Rat
Isotype IgG2c
Flow cytometry Yes
Storage temperature Cool
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo