Tlačiť

Anti-TNFRSF12A Mouse monoclonal antibody Biotin [clone: ITEM-4]

ABCAAB93554-50
Anti-TNFRSF12A Mouse monoclonal antibody Biotin [clone: ITEM-4]
Type: Primary
Antigen: TNFRSF12A (tumor necrosis factor receptor superfamily, member 12A)
Clonality: monoclonal
Clone: ITEM-4
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR