Keep my session open?
Ending In 
The session is expired
Vaša relácia uplynula. Pre Vašu bezpečnosť sme Vás odhlásili.
Chcete sa opäť prihlásiť?

Anti-TNFRSF12A Mouse monoclonal antibody PE (Phycoerythrin) [clone: ITEM-4]

Dodávateľ: Abcam
Type: Primary
Antigen: TNFRSF12A (tumor necrosis factor receptor superfamily, member 12A)
Clonality: monoclonal
Clone: ITEM-4
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse



 

OBJEDNAŤ

Rozšíriť všetko / Schovať všetko
Veľkosť Pk Číslo v katalógu VWR Množstvo
100 Tests 50 Tests ABCAAB93555-50
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

ŠPECIFIKÁCIE

Antigen symbol TNFRSF12A
Antigen name tumor necrosis factor receptor superfamily, member 12A
Reactivity Human, Mouse
Conjugation PE (Phycoerythrin)
Clonality Monoclonal
Antibody type Primary
Clone ITEM-4
Host Mouse
Isotype IgG2b
Flow cytometry Yes
Storage temperature Cool
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo