Keep my session open?
Ending In 
The session is expired
Vaša relácia uplynula. Pre Vašu bezpečnosť sme Vás odhlásili.
Chcete sa opäť prihlásiť?

Anti-DO-11.10 T Cell Receptor Mouse monoclonal antibody Biotin [clone: KJ1-26 (KJ126+KJI-26+KJI26)]

Dodávateľ: Abcam
Type: Primary
Antigen: DO-11.10 T Cell Receptor
Clonality: monoclonal
Clone: KJ1-26 (KJ126+KJI-26+KJI26)
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse



 

OBJEDNAŤ

Rozšíriť všetko / Schovať všetko
Veľkosť Pk Číslo v katalógu VWR Množstvo
100 µg 100 µG ABCAAB93556-100
Vytvoriť čiarové kódy Formát čiarového kódu: 
Vytvoriť katalóg v PDF

ŠPECIFIKÁCIE

Antigen name DO-11.10 T Cell Receptor
Reactivity Mouse
Conjugation Biotin
Clonality Monoclonal
Antibody type Primary
Clone KJ1-26 (KJ126+KJI-26+KJI26)
Host Mouse
Isotype IgG2a
Flow cytometry Yes
Storage temperature Cool
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo