Tlačiť

Anti-FASLG Mouse monoclonal antibody PE (Phycoerythrin) [clone: NOK-1]

ABCAAB93575-100
Anti-FASLG Mouse monoclonal antibody PE (Phycoerythrin) [clone: NOK-1]
Type: Primary
Antigen: FASLG (Fas ligand (TNF superfamily, member 6))
Clonality: monoclonal
Clone: NOK-1
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR