Tlačiť

Anti-PCNA Mouse monoclonal antibody PE (Phycoerythrin) [clone: PC10]

ABCAAB93576-100
Anti-PCNA Mouse monoclonal antibody PE (Phycoerythrin) [clone: PC10]
Type: Primary
Antigen: PCNA (proliferating cell nuclear antigen)
Clonality: monoclonal
Clone: PC10
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR