Tlačiť
Anti-Ly76 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: TER-119]
Dodávateľ: Abcam
ABCAAB93582-100EA 1047.71 EUR
ABCAAB93582-100
Anti-Ly76 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: TER-119]
Protilátky
Type: Primary
Antigen: Ly76
Clonality: monoclonal
Clone: TER-119
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse