Tlačiť

Anti-AICDA Rabbit Polyclonal Antibody

Dodávateľ: Abcam
ABCAAB93596-100EA 1068.17 EUR
ABCAAB93596-100
Anti-AICDA Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: AICDA (activation-induced cytidine deaminase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR