Tlačiť
Anti-TIMP3 Rabbit Polyclonal Antibody
Dodávateľ: Abcam
ABCAAB93637-100EA 1068.17 EUR
ABCAAB93637-100
Anti-TIMP3 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: TIMP3 (TIMP metallopeptidase inhibitor 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human