Tlačiť

Anti-NPHS2 Rabbit Polyclonal Antibody

Dodávateľ: Abcam
ABCAAB93650-100EA 857.03 EUR
ABCAAB93650-100
Anti-NPHS2 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: NPHS2 (nephrosis 2, idiopathic, steroid-resistant (podocin))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR