Tlačiť

Anti-MPO Rabbit monoclonal antibody [clone: SP72]

Dodávateľ: Abcam
ABCAAB93665-100EA 1059.47 EUR
ABCAAB93665-100
Anti-MPO Rabbit monoclonal antibody [clone: SP72]
Type: Primary
Antigen: MPO (myeloperoxidase)
Clonality: monoclonal
Clone: SP72
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR