Tlačiť

Anti-DNA Polymerase beta Rabbit polyclonal antibody unconjugated

ABCAAB93673-50
Anti-DNA Polymerase beta Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: DNA Polymerase beta
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR