Tlačiť

Anti-TUBG1 Rabbit Polyclonal Antibody

Dodávateľ: Abcam
ABCAAB93867-100EA 844.87 EUR
ABCAAB93867-100
Anti-TUBG1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: TUBG1 (tubulin, gamma 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR