Tlačiť

Anti-TNFAIP3 Rabbit Polyclonal Antibody

Dodávateľ: Abcam
ABCAAB93868-100EA 1065.85 EUR
ABCAAB93868-100
Anti-TNFAIP3 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: TNFAIP3 (tumor necrosis factor, alpha-induced protein 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR