Tlačiť

Anti-LMTK2 Goat polyclonal antibody

ABCAAB106835-100
Anti-LMTK2 Goat polyclonal antibody
Type: Primary
Antigen: LMTK2 (lemur tyrosine kinase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR