Tlačiť

Anti-FAS Goat polyclonal antibody

ABCAAB106839-100
Anti-FAS Goat polyclonal antibody
Type: Primary
Antigen: FAS (Fas cell surface death receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR