Tlačiť

Anti-HLA G Mouse monoclonal antibody [clone: MEM-G/1]

ABCAAB106841-50
Anti-HLA G Mouse monoclonal antibody [clone: MEM-G/1]
Type: Primary
Antigen: HLA G
Clonality: monoclonal
Clone: MEM-G/1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR